missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ I-Series Stackable Shakers: Stacking Kits

For use only with I-Series Stackable Shakers, Models I-26 and I-26R

Brand:  Eppendorf™ M1324-0600

 View more versions of this product

Product Code. 11692754

  • 9196.64 SEK / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket

Requires : Sticky Pad Platform (14-285-746) requires Sticky Pad (14-285-753, sold separately)
This item is not returnable. View return policy

Description

Description

  • Stacking kit enables two I-26 Series Stackable Shakers to be stacked vertically
  • Bases allow shaker(s) to be raised to suitable height

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Base Riser for three stack
10.16 cm
Short
4 in.
NBS I-Series Stackable Shakers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.