Learn More
Eppendorf™ Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ M1194-9908
 View more versions of this product
Product Code. 11385974
  • 12123.30 SEK / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
  • Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
  • Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dedicated Shaker Platform
18 in.
457 mm
18 in.
46 cm
5
18 x 18 in. (457 x 457 mm)
18 in.
46 cm
457 mm
I-2400, Innova 2100, Excella E24/E24R shaker