Learn More
Eppendorf™ Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
6465.76 SEK - 21294.84 SEK
Product Code | Brand | For Use With (Equipment) | No. of Positions | Length (Metric) Platform | Width (Metric) Platform | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | No. of Positions | Length (Metric) Platform | Width (Metric) Platform | Price | Quantity & Availability | ||
11688643
|
Eppendorf™
M1194-9920 |
Innova Model 2150 shaker flask | 8 | 61 cm | 46 cm |
12123.30 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11658623
|
Eppendorf™
M1190-9906 |
Innova 2000, Excella E1/E2, all I-Series shaker | 8 | 40.6 cm | 30.4 cm |
8257.90 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11336004
|
Eppendorf™
M1191-9910 |
Innova 2300, Excella E5/E10, I-2500/I-2500KC | 40 | 76 cm | 46 cm |
19713.54 SEK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11648623
|
Eppendorf™
M1190-9905 |
Innova 2000, Excella E1/E2, all I-Series shaker | 12 | 40.6 cm | 30.4 cm |
9417.52 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11335974
|
Eppendorf™
M1194-9903 |
Innova 2100, Excella E5, I-2500/I-2500KC | 64 | 46 cm | 46 cm |
14864.22 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11365974
|
Eppendorf™
M1194-9906 |
Innova 2100, Excella E5, I-2500/I-2500KC | 16 | 46 cm | 46 cm |
12650.40 SEK
Each |
In Stock
Log in to see stock. |
||
11355974
|
Eppendorf™
M1194-9905 |
Innova 2100, Excella E5, I-2500/I-2500KC | 25 | 46 cm | 46 cm |
13177.50 SEK
Each |
In Stock
Log in to see stock. |
||
11345974
|
Eppendorf™
M1194-9904 |
I-2400, Innova 2100, Excella E24/E24R shaker | 34 | 46 cm | 46 cm |
12650.40 SEK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11608633
|
Eppendorf™
M1190-9915 |
Innova 2000, Excella E1/E2, all I-Series shaker | 20 | 33 cm | 28 cm |
6465.76 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11618633
|
Eppendorf™
M1190-9916 |
Innova 2000, Excella E1/E2, all I-Series shaker | 12 | 33 cm | 28 cm |
6465.76 SEK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11638633
|
Eppendorf™
M1190-9918 |
Innova 2000, Excella E1/E2, all I-Series shaker | 6 | 33 cm | 28 cm |
6867.36 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11628633
|
Eppendorf™
M1190-9917 |
Innova 2000, Excella E1/E2, all I-Series shaker | 8 | 33 cm | 28 cm |
6867.36 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11385974
|
Eppendorf™
M1194-9908 |
I-2400, Innova 2100, Excella E24/E24R shaker | 5 | 46 cm | 46 cm |
12123.30 SEK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11375974
|
Eppendorf™
M1194-9907 |
I-2400, Innova 2100, Excella E24/E24R shaker | 9 | 46 cm | 46 cm |
12123.30 SEK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
11326004
|
Eppendorf™
M1191-9909 |
Innova 2300, Excella E5/E10, I-2500/I-2500KC | 65 | 76 cm | 46 cm |
21294.84 SEK
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
Description
- Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
- Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Dedicated Shaker Platform |