Learn More
Eppendorf™ Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
6465.76 SEK - 21294.84 SEK
Specifications
Product Type Dedicated Shaker Platform
View More Specs
Products 15
Product Code Brand For Use With (Equipment) No. of Positions Length (Metric) Platform Width (Metric) Platform Price Quantity & Availability  
Product Code Brand For Use With (Equipment) No. of Positions Length (Metric) Platform Width (Metric) Platform Price Quantity & Availability  
11688643
View Documents
Eppendorf™
M1194-9920
Innova Model 2150 shaker flask 8 61 cm 46 cm
12123.30 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11658623
View Documents
Eppendorf™
M1190-9906
Innova 2000, Excella E1/E2, all I-Series shaker 8 40.6 cm 30.4 cm
8257.90 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11336004
View Documents
Eppendorf™
M1191-9910
Innova 2300, Excella E5/E10, I-2500/I-2500KC 40 76 cm 46 cm
19713.54 SEK
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11648623
View Documents
Eppendorf™
M1190-9905
Innova 2000, Excella E1/E2, all I-Series shaker 12 40.6 cm 30.4 cm
9417.52 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11335974
View Documents
Eppendorf™
M1194-9903
Innova 2100, Excella E5, I-2500/I-2500KC 64 46 cm 46 cm
14864.22 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11365974
View Documents
Eppendorf™
M1194-9906
Innova 2100, Excella E5, I-2500/I-2500KC 16 46 cm 46 cm
12650.40 SEK
Each
In Stock
to see stock.
Add to basket
11355974
View Documents
Eppendorf™
M1194-9905
Innova 2100, Excella E5, I-2500/I-2500KC 25 46 cm 46 cm
13177.50 SEK
Each
In Stock
to see stock.
Add to basket
11345974
View Documents
Eppendorf™
M1194-9904
I-2400, Innova 2100, Excella E24/E24R shaker 34 46 cm 46 cm
12650.40 SEK
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11608633
View Documents
Eppendorf™
M1190-9915
Innova 2000, Excella E1/E2, all I-Series shaker 20 33 cm 28 cm
6465.76 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11618633
View accessory products View Documents
Eppendorf™
M1190-9916
Innova 2000, Excella E1/E2, all I-Series shaker 12 33 cm 28 cm
6465.76 SEK
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11638633
View Documents
Eppendorf™
M1190-9918
Innova 2000, Excella E1/E2, all I-Series shaker 6 33 cm 28 cm
6867.36 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11628633
View Documents
Eppendorf™
M1190-9917
Innova 2000, Excella E1/E2, all I-Series shaker 8 33 cm 28 cm
6867.36 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
11385974
View accessory products View Documents
Eppendorf™
M1194-9908
I-2400, Innova 2100, Excella E24/E24R shaker 5 46 cm 46 cm
12123.30 SEK
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11375974
View accessory products View Documents
Eppendorf™
M1194-9907
I-2400, Innova 2100, Excella E24/E24R shaker 9 46 cm 46 cm
12123.30 SEK
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11326004
View Documents
Eppendorf™
M1191-9909
Innova 2300, Excella E5/E10, I-2500/I-2500KC 65 76 cm 46 cm
21294.84 SEK
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
Description
Description
  • Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
  • Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Specifications
Dedicated Shaker Platform