Learn More
Eppendorf™ Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ M1190-9915
 View more versions of this product
Product Code. 11608633
  • 6465.76 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
  • Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
  • Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dedicated Shaker Platform
13 x 11 in. (330 x 279 mm)
13 in.
33 cm
11 in.
28 cm
20
Stainless Steel
13 in.
330 mm
11 in.
279 mm
Innova 2000, Excella E1/E2, all I-Series shaker