Learn More
Eppendorf™ Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand: Eppendorf™ M1190-9917
This item is not returnable.
View return policy
Description
- Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
- Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Dedicated Shaker Platform | |
13 x 11 in. (330 x 279 mm) | |
13 in. | |
33 cm | |
11 in. | |
28 cm | |
8 |
Stainless Steel | |
13 in. | |
330 mm | |
11 in. | |
279 mm | |
Innova 2000, Excella E1/E2, all I-Series shaker |