Learn More
Eppendorf™ Scientific Shaker Utility Trays
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ M1194-9910
 View more versions of this product
Product Code. 11305974
  • 9753.86 SEK / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Description
  • For shaking vessels without fasteners
  • Trays have nonskid rubber mat
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Utility Tray
18 in.
46 cm
18 in.
46 cm
Non-skid Rubber surface
18 x 18 in. (46 x 46 cm)
18 in.
46 cm
46 cm
Innova 2100, Excella E24/E24R, I-24/!-24R