Learn More
Eppendorf™ Scientific Shaker Utility Trays
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ AG2-00
 View more versions of this product
Product Code. 11678603
  • 7203.70 SEK / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Description
  • For shaking vessels without fasteners
  • Trays have nonskid rubber mat
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Utility Tray
33 cm
28 cm
Non-skid Rubber surface
13 in.
11 in.
Innova 2000, Excella E1/E2