Learn More
Eppendorf™ Scientific Shaker Utility Trays
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand:  Eppendorf™ M1194-9922
 View more versions of this product
Product Code. 11698643
  • 11385.36 SEK / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Description
  • For shaking vessels without fasteners
  • Trays have nonskid rubber mat
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Utility Tray (for low-speed applications)
18 in.
457 mm
24 in.
610 mm
Innova Model 2150 shaker
18 x 24 in. (457 x 610 mm)
24 in.
61 cm
18 in.
46 cm
Non-skid Rubber surface