Learn More
Eppendorf™ Scientific Shaker Utility Trays
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand: Eppendorf™ M1194-9922
This item is not returnable.
View return policy
Description
- For shaking vessels without fasteners
- Trays have nonskid rubber mat
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Utility Tray (for low-speed applications) | |
18 in. | |
457 mm | |
24 in. | |
610 mm | |
Innova Model 2150 shaker |
18 x 24 in. (457 x 610 mm) | |
24 in. | |
61 cm | |
18 in. | |
46 cm | |
Non-skid Rubber surface |