missing translation for 'onlineSavingsMsg'
Learn More

NITD008, MedChemExpress

MedChemExpress NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.

Encompass_Preferred

POA - 5306.16 SEK

Specifications

Chemical Name or Material NITD008
CAS 1044589-82-3
Color Gray
Purity Grade Notes Research
Molecular Formula C13H14N4O4
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475448
Encompass_Preferred
View Documents
Medchem Express
HY-12957
10 mg
5306.16 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16461319
Encompass_Preferred
View Documents
Medchem Express
HY-12957
1 mg
POA
1mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16418167
Encompass_Preferred
View Documents
Medchem Express
HY-12957
5 mg
3376.65 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.
Specifications

Specifications

NITD008
Gray
C13H14N4O4
DMSO : ≥ 50 mg/mL (172.25 mM)
290.27
96.2%
Solid
1044589-82-3
Research
7-Deaza-2'-C-acetylene-adenosine
O[C@@]1(C#C)[C@H](N2C=CC3=C(N)N=CN=C32)O[C@H](CO)[C@H]1O
290.27
Research
COVID-19-anti-virus
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Certificates

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
NITD008, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.