Learn More
NITD008, MedChemExpress
MedChemExpress NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.
Brand: Medchem Express HY-12957
Description
NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.Specifications
NITD008 | |
Gray | |
C13H14N4O4 | |
7-Deaza-2'-C-acetylene-adenosine | |
O[C@@]1(C#C)[C@H](N2C=CC3=C(N)N=CN=C32)O[C@H](CO)[C@H]1O | |
290.27 | |
Research | |
COVID-19-anti-virus |
1044589-82-3 | |
Research | |
5 mg | |
DMSO : ≥ 50 mg/mL (172.25 mM) | |
290.27 | |
96.2% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only