missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLGN4Y Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13489-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody can recognize both of NLGN4Y and NLGN4X at 72 kDa mature form.
Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 ).Specifications
NLGN4Y | |
Polyclonal | |
Unconjugated | |
NLGN4Y | |
KIAA0951, neuroligin 4, Y linked, Neuroligin Y, NLGN4Y | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22829, 57502 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N0W4, Q8NFZ3 | |
NLGN4X, NLGN4Y | |
NLGN4Y Fusion Protein Ag4311 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction