Learn More
Masterflex™ Non-DEHP Lab Tubing
Use this tubing for transfer of liquids, gases and inorganic chemicals found in lab, paint, pigment and soap dispensing applications. Cole-Parmer™ Non-DEHP Lab Tubing is ideal for analytical instruments and general laboratory needs.
Brand: Masterflex™ 95505-02
Description
- Nontoxic DEHP-free formulation
- Flexible, small bend radii
- Transparent; allows for visual inspection and flow monitoring
- Certifications: FDA compliant, Proposition 65, REACH
Specifications
Autoclavable | |
4 mm | |
0.8 mm | |
1 Pack | |
Transparent | |
32 psi |
2.4 mm | |
15.25 m | |
-50°C | |
Sterile | |
Flexible | |
73°C |
Product Suggestions
Customers who viewed this item also viewed.