Learn More
GPE Scientific™ Norell™ Thin-Wall Transparent Fluoropolymer FEP Tubing
Manufactured from special virgin grade thermoplastic FEP fluoropolymer
Brand: GPE Scientific™ TWT-7
Additional Details : Weight : 1.00000kg
Description
- Unlike PTFE tubing, this material can be heat sealed
- Heat sealing can offer extremely large possibilities for packaging and/or storage of samples (corrosives, liquids, solids, etc.)
- Fluoropolymer tubing can also be used where snug and tight fit over glass or metal tubing is required
Specifications
7 mm | |
7.68 mm | |
where snug and tight fit over glass or metal tubing is required |
FEP | |
305 mm | |
4/Pk. |