Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
Brand: Thermo Scientific™ 4021
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
- Available with a molded grid
- Performs well in automatic dispensers
- Contact dish useful for sampling in hospital environments as well as in food science and the pharmaceutical industry
- Compatible with automated systems
Specifications
Vented, without stack ring | |
79 sq. cm | |
Petri Dish | |
Yes |
Flat | |
Square | |
1 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.