Learn More
Kern Sohn OIML Class E1 Weight Set, Aluminum Case
Weight set constructed of polished stainless steel and housed in an aluminum case.
Brand:  Kern Sohn 304-046
Encompass_Preferred
 View more versions of this product
Product Code. 17301843
  • 17900.76 SEK / 1 set
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Description
  • Set of weights
  • Polished stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
E1
Stainless Steel
Aluminum
1 to 200 g