Learn More
Kern Sohn OIML Class E1 Weight Set, Aluminum Case
Weight set constructed of polished stainless steel and housed in an aluminum case.
10920.59 SEK - 89166.02 SEK
Specifications
Classification E1
Material Stainless Steel
Case Material Aluminum
View More Specs
Products 16
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
17301823
Encompass_Preferred
View Documents
Kern Sohn
303-026
1 mg to 50 g
19814.67 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17321823
Encompass_Preferred
View Documents
Kern Sohn
303-036
1 mg to 100 g
21503.43 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17341823
Encompass_Preferred
View Documents
Kern Sohn
303-046
1 mg to 200 g
25556.43 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17361823
Encompass_Preferred
View Documents
Kern Sohn
303-056
1 mg to 500 g
29384.26 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17381823
Encompass_Preferred
View Documents
Kern Sohn
303-066
1 mg to 1 kg
37602.84 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17301833
Encompass_Preferred
View Documents
Kern Sohn
303-076
1 mg to 2 kg
51450.60 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17321833
Encompass_Preferred
View Documents
Kern Sohn
303-086
1 mg to 5 kg
66311.61 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17341833
Encompass_Preferred
View Documents
Kern Sohn
303-096
1 mg to 10 kg
89166.02 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17361833
Encompass_Preferred
View Documents
Kern Sohn
304-026
1 to 50 g
10920.59 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17381833
Encompass_Preferred
View Documents
Kern Sohn
304-036
1 to 100 g
12834.51 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17301843
Encompass_Preferred
View Documents
Kern Sohn
304-046
1 to 200 g
17900.76 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17321843
Encompass_Preferred
View Documents
Kern Sohn
304-056
1 to 500 g
21503.43 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17341843
Encompass_Preferred
View Documents
Kern Sohn
304-066
1 g to 1 kg
26344.51 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17361843
Encompass_Preferred
View Documents
Kern Sohn
304-076
1 g to 2 kg
42443.93 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17381843
Encompass_Preferred
View Documents
Kern Sohn
304-086
1 g to 5 kg
55391.02 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17301853
Encompass_Preferred
View Documents
Kern Sohn
304-096
1 g to 10 kg
76669.27 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
Description
Description
  • Set of weights
  • Polished stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
E1
Stainless Steel
Aluminum