Learn More
Kern Sohn OIML Class E2 Weight Set, Plastic Case
Weight set constructed of polished stainless steel and housed in a plastic case.
Brand: Kern Sohn 313-074
This item is not returnable.
View return policy
Description
- Set of weights
- Polished stainless steel
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
E2 | |
Stainless Steel | |
Plastic | |
1 mg to 2 kg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.