Learn More
Kern Sohn OIML Class E2 Weight Set, Plastic Case
Weight set constructed of polished stainless steel and housed in a plastic case.
6466.30 SEK - 34330.15 SEK
Specifications
Classification E2
Material Stainless Steel
Case Material Plastic
View More Specs
Products 14
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
17371883
Encompass_Preferred
View Documents
Kern Sohn
313-024
1 mg to 50 g
10346.07 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17311893
Encompass_Preferred
View Documents
Kern Sohn
313-034
1 mg to 100 g
11286.63 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17351893
Encompass_Preferred
View Documents
Kern Sohn
313-044
1 mg to 200 g
14343.42 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17311903
Encompass_Preferred
View Documents
Kern Sohn
313-054
1 mg to 500 g
15989.39 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17341903
Encompass_Preferred
View Documents
Kern Sohn
313-064
1 mg to 1 kg
19046.18 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17381903
Encompass_Preferred
View Documents
Kern Sohn
313-074
1 mg to 2 kg
26335.46 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17341913
Encompass_Preferred
View Documents
Kern Sohn
313-084
1 mg to 5 kg
34330.15 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17301923
Encompass_Preferred
View Documents
Kern Sohn
314-024
1 to 50 g
6466.30 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17331923
Encompass_Preferred
View Documents
Kern Sohn
314-034
1 to 100 g
7289.28 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17361923
Encompass_Preferred
View Documents
Kern Sohn
314-044
1 to 200 g
9758.23 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17381923
Encompass_Preferred
View Documents
Kern Sohn
314-054
1 to 500 g
12579.88 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17311933
Encompass_Preferred
View Documents
Kern Sohn
314-064
1 g to 1 kg
15166.40 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17341933
Encompass_Preferred
View Documents
Kern Sohn
314-074
1 g to 2 kg
22455.68 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17361933
Encompass_Preferred
View Documents
Kern Sohn
314-084
1 g to 5 kg
28334.13 SEK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
  • Set of weights
  • Polished stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
E2
Stainless Steel
Plastic
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.