Learn More
Ohaus™ OIML F1 Weight Set
Adjusted to Meet or Exceed OIML F1 Tolerances
3410.00 SEK - 27100.00 SEK
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16672462
|
Ohaus™
80780408 |
500 mg to 1 mg |
3410.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16682462
|
Ohaus™
80780410 |
50 g to 1 mg |
6770.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16692462
|
Ohaus™
80780411 |
50 g to 1 g |
4205.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
16602472
|
Ohaus™
80780412 |
100 g to 1 mg |
7505.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16612472
|
Ohaus™
80780413 |
100 g to 1 g |
5080.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16622472
|
Ohaus™
80780414 |
2 x 200 g to 1 mg |
9695.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16632472
|
Ohaus™
80780415 |
2 x 200 g to 1 g |
6465.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16642472
|
Ohaus™
80780418 |
500 g to 1 mg |
10370.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
16652472
|
Ohaus™
80780419 |
500 g to 1 g |
7685.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16662472
|
Ohaus™
80780420 |
1 kg to 1 mg |
17045.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16672472
|
Ohaus™
80780421 |
1 kg to 1 g |
14675.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16682472
|
Ohaus™
80780422 |
2 x 2 kg to 1 mg |
22250.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16692472
|
Ohaus™
80780423 |
2 x 2 kg to 1 g |
20830.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16602482
|
Ohaus™
80780426 |
5 kg to 1 mg |
27100.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16612482
|
Ohaus™
80780427 |
5 kg to 1 g |
23905.00 SEK
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
Description
- Weights Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free to protect your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
Protective Case |