Learn More
Tocris Bioscience™ OXF BD 02
Selective BRD4(1) inhibitor
Brand: Tocris Bioscience™ 4928/50
Additional Details : CAS Number : 1429129-68-9 Weight : 0.00970kg
Description
OXF BD 02 is a selective inhibitor of the first bromodomain of BRD4 (BRD4(1)) (IC50 = 382 nM). Exhibits 2-3-fold selectivity for BRD4(1) over the CBP bromodomain and has little affinity for a range of other bromodomains. Reduces viability of lung adenocarcinoma cell lines and attenuates proliferation of MV-4-11 leukemia cells. Cell permeable.Specifications
>98% | |
OXF BD 02 | |
OXF BD 02 |
50 mg | |
295.33 |
For Research Use Only