missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OXF BD 02
Selective BRD4(1) inhibitor
1727.82 SEK - 6113.23 SEK
Specifications
Purity | >98% |
---|---|
Product Type | OXF BD 02 |
Molecular Weight (g/mol) | 295.33 |
Inhibitors | OXF BD 02 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722459
|
Tocris Bioscience™
4928/50 |
50 mg |
6113.23 SEK
50mg |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18791809
|
Tocris Bioscience™
4928/10 |
10 mg |
1727.82 SEK
10mg |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Selective inhibitor of the first bromodomain of BRD4 (BRD4(1)) (IC50 = 382 nM). Exhibits 2-3-fold selectivity for BRD4(1) over the CBP bromodomain and has little affinity for a range of other bromodomains. Reduces viability of lung adenocarcinoma cell lines and attenuates proliferation of MV-4-11 leukemia cells. Cell permeable.Specifications
>98% | |
OXF BD 02 | |
295.33 | |
OXF BD 02 |
For Research Use Only