Learn More
PDE-9 inhibitor, MedChemExpress
MedChemExpress PDE-9 inhibitor is useful for neurodegenerative diseases.
Brand:  Medchem Express HY-50865
Encompass_Preferred
 View more versions of this product
Product Code. 16468118
  • 5772.98 SEK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
PDE-9 inhibitor is useful for neurodegenerative diseases.
Specifications
Specifications
PDE-9 inhibitor
1082743-70-1
10 mg
C22H27N5O2
DMSO : ≥ 100 mg/mL (254.14 mM)
393.48
99.83%
Solid
White
Research
Neuroscience-Neurodegeneration
O=C1NC([C@H]2[C@@H](CN(C2)CC3=CC=CC=C3)C)=NC4=C1C=NN4C5CCOCC5
393.48
Research
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PDE-9 inhibitor, MedChemExpress