Learn More
PDE-9 inhibitor, MedChemExpress
MedChemExpress PDE-9 inhibitor is useful for neurodegenerative diseases.

3859.03 SEK - 15918.50 SEK
Specifications
Chemical Name or Material | PDE-9 inhibitor |
---|---|
Physical Form | Solid |
CAS | 1082743-70-1 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16470578
![]() |
Medchem Express
HY-50865 |
5 mg |
3859.03 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468118
![]() |
Medchem Express
HY-50865 |
10 mg |
5772.98 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444037
![]() |
Medchem Express
HY-50865 |
50 mg |
15918.50 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE-9 inhibitor is useful for neurodegenerative diseases.Specifications
PDE-9 inhibitor | |
1082743-70-1 | |
Research | |
Neuroscience-Neurodegeneration | |
O=C1NC([C@H]2[C@@H](CN(C2)CC3=CC=CC=C3)C)=NC4=C1C=NN4C5CCOCC5 | |
393.48 | |
Research |
Solid | |
White | |
C22H27N5O2 | |
DMSO : ≥ 100 mg/mL (254.14 mM) | |
393.48 | |
99.83% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.