Learn More
Gilac™ Perforated Crate
Perforated Crate is a stacking perforated crate ideal for storing delicate foodstuffs such as fruit and vegetables; it provides ventilation and is highly resistant to intensive use.
Brand:  Gilac™ G179721
 View more versions of this product
Product Code. 17998781
  • 501.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Stacking perforated crate.
  • Ideal for storing all products requiring air circulation.
  • Highly resistant for intensive use.
  • Ideal for storing fruit and vegetables.
  • Easy to grab hold of thanks to practical ergonomic integrated handles.
  • Solid container lids are also suitable for perforated containers.
  • Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Specifications
15.9 gal.
None
HDPE
Crate
600 x 400 x 300 mm
23.6 in.
15.7 in.
11.8 in.
60 L
White
Rectangular
Yes
600 mm
400 mm
300 mm