missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether 30-40°C, Extra Pure, SLR, Fisher Chemical™
234.00 SEK - 6120.00 SEK
Chemical Identifiers
CAS | 64742-49-0 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.178 |
MDL Number | MFCD00081849 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
PubChem CID | 7892 |
SMILES | CCCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11488712
|
Fisher Chemical
P/1400/08 |
500 mL | Amber Glass Bottle | N/A | |||||
10635692
|
Fisher Chemical
P/1400/17 |
2.5 L | Amber Glass Bottle | N/A | |||||
11904071
|
Fisher Chemical
P/1400/MS21 |
5 L | Aluminum Can | N/A | |||||
10294150
|
Fisher Chemical
P/1400/25 |
25 L | Metal Can |
6120.00 SEK
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
64742-49-0 | |
86.178 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 |
C6H14 | |
MFCD00081849 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C |
Specifications
-130°C | |
30°C | |
C6H14 | |
514mmHg 25 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
Petroleum ether | |
7892 |
64742-49-0 | |
Liquid | |
MFCD00081849 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
CCCC(C)C | |
86.178 | |
72.15g/mol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title