missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Straight Pipe Systems

The SymLine™ pipe system provides connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!

1145.00 SEK - 1535.00 SEK

Specifications

Diameter (Metric) Outer 32 mm
Material HDPE-EC
View More Specs

Products 3
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
14776901
View Documents
SymLine™
106435
300 mm
1145.00 SEK
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
14786901
View Documents
SymLine™
106436
900 mm
1220.00 SEK
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
14796901
View Documents
SymLine™
106437
1200 mm
1535.00 SEK
Each
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
 
Description

Description

  • Straight pipes are available in multiple standard lengths
  • A wide range of connectors, branches and manifolds allows the pipe system to be connected precisely to the conditions in your laboratory
Specifications

Specifications

32 mm
HDPE-EC
Videos
SDS
Documents

Documents

Product Certifications