Learn More
Microspec™ Pipette Tips
Available as sterile or non-sterile
Brand:  Microspec™ AE3
 View more versions of this product
Product Code. 16605472
  • 249.00 SEK / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available as sterile or non-sterile
  • In bulk bags or in grip seal bags
  • Non-filtered
Specifications
Specifications
200 μL
Non-sterile
Not Autoclavable
1000
Non-filtered
Blue
Bulk Bag
Single-use