Learn More
Microspec™ Pipette Tips
Available as sterile or non-sterile
249.00 SEK - 980.00 SEK
Specifications
Filtered | Non-filtered |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
16685462
|
Microspec™
AE1 |
10 μL | Non-sterile |
396.00 SEK
Pack of 1000 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
16605472
|
Microspec™
AE3 |
200 μL | Non-sterile |
249.00 SEK
Pack of 1000 |
In Stock
Log in to see stock. |
||||
16615472
|
Microspec™
AE5 |
1000 μL | Non-sterile |
267.00 SEK
Pack of 1000 |
In Stock
Log in to see stock. |
||||
16625472
|
Microspec™
AE9 |
5 mL | Non-sterile |
535.00 SEK
Pack of 300 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
16695462
|
Microspec™
AE10 |
5 mL | Sterile |
980.00 SEK
Pack of 300 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
Description
- Available as sterile or non-sterile
- In bulk bags or in grip seal bags
- Non-filtered
Specifications
Non-filtered | |
Not Autoclavable | |
Single-use |