Learn More
Eppendorf™ Platform for Innova™ 2050 Shaker
This dedicated platform fits shaker model 2050.
Brand: Eppendorf™ M1190-9901
This item is not returnable.
View return policy
Description
- Holds Erlenmeyer flasks
- Dedicated platform
- Made of stainless steel
Specifications
10 mL Erlenmeyer Flasks | |
41 x 31 cm | |
86 |
Stainless Steel | |
Innova™ 2000 Shaker |