Learn More
Eppendorf™ Platforms for Innova™ 2150 Shaker
This dedicated platform fits shaker model 2150.
Brand:  Eppendorf™ M1194-9915
 View more versions of this product
Product Code. 11638643
  • 18870.18 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Holds Erlenmeyer flasks
  • Dedicated platform
  • Made of phenolic resin
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL Erlenmeyer Flasks
61 x 45.7 cm
80
Phenolic Resin
Innova™ 2300/43/43R and Excella™ E10/E25/E25R Shakers