Learn More
Eppendorf™ Platforms for Innova™ 2150 Shaker
This dedicated platform fits shaker model 2150.
Brand:  Eppendorf™ M1194-9917
 View more versions of this product
Product Code. 11658643
  • 16550.94 SEK / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Description
  • Holds Erlenmeyer flasks
  • Dedicated platform
  • Made of phenolic resin
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL Erlenmeyer Flasks
61 x 45.7 cm
35
Phenolic Resin
Innova™ 2300/43/43R and Excella™ E10/E25/E25R Shakers