Learn More
Eppendorf™ Platforms for Innova™ 2150 Shaker
This dedicated platform fits shaker model 2150.
Brand:  Eppendorf™ M1194-9919
 View more versions of this product
Product Code. 11678643
  • 14653.38 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
  • Holds Erlenmeyer flasks
  • Dedicated platform
  • Made of phenolic resin
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 L Erlenmeyer Flasks
61 x 45.7 cm
12
Phenolic Resin
Innova™ 2300/43/43R and Excella™ E10/E25/E25R Shakers