Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
Brand:  Eppendorf™ S44I040004
Additional Details : Weight : 12.31000kg
 View more versions of this product
Product Code. 15804065
  • 15960.00 SEK / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
  • Dedicated platform
Specifications
Specifications
500 mL Erlenmeyer Flasks
Innova™ S44i
39
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Ofertas especiales
Ofertas especiales
Servicio Técnico