Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
16550.94 SEK - 21294.84 SEK
Specifications
For Use With (Equipment) Innova™ S44i
View More Specs
Products 8
Product Code Brand No. of Positions Holds Price Quantity & Availability  
Product Code Brand No. of Positions Holds Price Quantity & Availability  
15884055
View accessory products View Documents
Eppendorf™
S44I040002
86 125 mL Erlenmeyer Flasks
21294.84 SEK
Each
In Stock
to see stock.
Add to basket
15894055
View accessory products View Documents
Eppendorf™
S44I040003
53 250 mL Erlenmeyer Flasks
19713.54 SEK
Each
In Stock
to see stock.
Add to basket
15804065
View accessory products View Documents
Eppendorf™
S44I040004
39 500 mL Erlenmeyer Flasks
18448.50 SEK
Each
In Stock
to see stock.
Add to basket
15814065
View accessory products View Documents
Eppendorf™
S44I040005
15 1 L Erlenmeyer Flasks
17499.72 SEK
Each
In Stock
to see stock.
Add to basket
15824065
View accessory products View Documents
Eppendorf™
S44I040006
15 2 L Erlenmeyer Flasks
16550.94 SEK
Each
In Stock
to see stock.
Add to basket
15854065
View accessory products View Documents
Eppendorf™
S44I040010
15 2 L Lauber Tomson Flasks
20240.64 SEK
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
15834065
View accessory products View Documents
Eppendorf™
S44I040007
8 2.8 L Fernbach Flasks
17499.72 SEK
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
15844065
View accessory products View Documents
Eppendorf™
S44I040008
8 4 L Erlenmeyer Flasks
16550.94 SEK
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
  • Dedicated platform
Specifications
Specifications
Innova™ S44i