Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
Brand:  Eppendorf™ S44I040006
 View more versions of this product
Product Code. 15824065
  • 16550.94 SEK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
  • Dedicated platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 L Erlenmeyer Flasks
Innova™ S44i
15
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products