Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
Brand: Eppendorf™ S44I040006
Additional Details : Weight : 13.94000kg
Description
- Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
- Dedicated platform
Specifications
2 L Erlenmeyer Flasks | |
Innova™ S44i | |
15 |