Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
Brand:  Eppendorf™ S44I040008
 View more versions of this product
Product Code. 15844065
  • 16550.94 SEK / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
  • Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
  • Dedicated platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 L Erlenmeyer Flasks
Innova™ S44i
8