Learn More
Eurofins DiscoverX Plk1 Protein, Active
Active, N-terminal 6His tagged, recombinant, human Plk1, for use in Kinase Assays.
5796.39 SEK - 44240.30 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 69.3 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17300704
|
Eurofins DiscoverX
14-777 |
10 μg |
5796.39 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17310704
|
Eurofins DiscoverX
14-777M |
250 μg |
44240.30 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Polo-like kinases (PLKs) are a family of serine/threonine kinases related to the polo gene product of Drosophila melanogaster, originally identified from a mitotic mutant that displayed abnormal spindle poles. PLKs play essential roles in several stages of mitosis and cytokinesis with a dynamic pattern of localization to centrosomes, kinetochores and central spindle structures during the cell cycle. PLK1 has been shown to be overexpressed in many human tumours, such as non-small cell lung cancer, squamous cell carcinomas of the head and neck, melanomas, oropharyngeal carcinomas, and ovarian and endometrial carcinomas.
GenBank NM_005030
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Full-length | |
Plk1, active |
69.3 kDa | |
Recombinant | |
Recombinant protein expressed in Sf21 insect cells | |
≥72% |