Learn More
Memmert™ Plug-In Tube Extension for UF160plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ UF160plusB29718
Product Code. 17509055
  • 549.00 SEK / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
UF160plus Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm
Product Title
Memmert™ Plug-In Tube Extension for UF160plus, Straight > For Use With: UF160plus