Learn More
Brady™ GlobalMark™ Polyester Cold Temperature Tape
GlobalMark™ Polyester Cold Temperature Tape permanently adheres to surfaces at temperatures as low as -10 degrees F. Great for use in any cold storage warehouse, cooler or freezer. Not recommended for plotter cutting.
Brand:  Brady™ 076691
 View more versions of this product
Product Code. 11549263
  • 8655.00 SEK / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • GlobalMark™ cold temperature tape permanently adheres to surfaces at temperatures as low as -10°F.
  • Great for use in any cold storage warehouse, cooler or freezer.
  • Not recommended for plotter cutting.
Specifications
Specifications
GlobalMark Cold Temperature Tape
White
Roll
Thermal Transfer
General Identification, Laboratory labelling, Safety Identification, Temperature indicating labelling
20 Gardner Units
1 Year
1 Roll/Box
-40°F to +212°F
0.003 in.
30.48 m x 107.95 mm
30.48 m
107.95 mm
Permanent Acrylic
Gloss
Polyester
Labels
GlobalMark Printer
1
-40°C
1 Ea.
-40°C to +100°C
0.076 mm
100 ft.
4.25 in.