Learn More
RETSCH Polystyrene Mixing Beakers
Brand:  RETSCH 22.041.0004
2546.00 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 11914362
  • 2680.00 SEK / Each
Estimated Shipment: 06-11-2024
to see stock.
Specifications
Mixing Beaker
56 mL
For Mixer Mill MM 400
Polystyrene