Learn More
Hach Lange™ Portable Turbidimeter
Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement.
Brand: Hach Lange™ 2100QIS01
This item is not returnable.
View return policy
Description
- With easy-to-follow on-screen instructions, manuals or memorization are not needed for routine calibrations.
- The Rapidly Settling Turbidity mode allows for accurate measurements even with the most difficult-to-measure settling samples.
- The rugged design makes this the instrument of choice for the toughest environments.
Specifications
LED | |
Portable Turbidimeter | |
±2% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Hach Lange™ Portable Turbidimeter > Accuracy: ±2%
Spot an opportunity for improvement?Share a Content Correction