Learn More
Labcold™ Powder Coated Steel Pharmacy Refrigerators
158785.00 SEK
Products 1
Product Code Brand Dimensions (DxWxH) Exterior Price Quantity & Availability  
Product Code Brand Dimensions (DxWxH) Exterior Price Quantity & Availability  
12928614
View Documents
Labcold™
RPFR21043
820 x 720 x 1980mm
158785.00 SEK
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
Description
Description
  • Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation