Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
1526.55 SEK - 8106.13 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11327151
|
Thermo Scientific™
09.4000 |
Double cartridge pretreatment system (includes 5µm filter with carbon cartridge and a hardness stabilizer cartridge) |
8106.13 SEK
Each |
In Stock
Log in to see stock. |
|||||
11766259
|
Thermo Scientific™
D8830 |
Barnstead Pre-Filter MQ Cartridge, 0.5 Micron |
2201.08 SEK
Each |
Estimated Shipment: 20-09-2024
Log in to see stock. |
|||||
15322178
|
Thermo Scientific™
D63111 |
Cellulose Acetate Prefilter, 5 Micron, for RoPure Reverse Osmosis Water Systems |
2047.24 SEK
1 unit |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
12756208
|
Thermo Scientific™
06.5204 |
5µm filter with hardness stabilizer |
1857.90 SEK
Each |
In Stock
Log in to see stock. |
|||||
11387101
|
Thermo Scientific™
06.5452 |
10 ft. (25.4 cm) Hardness stabilizing cartridge. For use with 09.4000 |
1857.90 SEK
Each |
In Stock
Log in to see stock. |
|||||
11367151
|
Thermo Scientific™
09.4003 |
Pretreatment filter for the Smart2Pure system. Includes filter housing and 1µm filter |
2153.74 SEK
Each |
In Stock
Log in to see stock. |
|||||
12818111
|
Thermo Scientific™
50134022 |
5µm filter with carbon with 10 ft. filter housing |
1526.55 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.