Learn More
Fisherbrand™ PTFE Stopper, Flask with Handle
Fisherbrand™ PTFE Stopper, Flask with Handle are for use with volumetric flasks and similar containers.
Brand:  Fisherbrand™ 010.4.38
 View more versions of this product
Product Code. 16235971
Description
Description
  • Solid Construction
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
Fisherbrand™ PTFE Stopper, Flask with Handle, Black Handle
1
-200°C to +280°C
PTFE
Standard Taper Ground Glass Joints
38 (US)
Black
Product Title
Fisherbrand™ PTFE Stopper, Flask with Handle > Size (US): 38; Handle Color: Black