Learn More
Quickfit™ Borosilicate Glass Round Bottom Flask with Septum Side Neck
Designed for semi-micro and micro scale preparations.
Brand:  Quickfit FR25/1S/SEP
 View more versions of this product
Product Code. 15527870
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • One vertical centre neck having precision-ground Quickfit™ socket to accept Quickfit™ cone
  • One angled side neck designed to take a rubber septum seal to allow small quantities of liquid to be handled easily and safely using a syringe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
Designed for semi-micro and micro scale preparations
Septum side neck
Round
Borosilicate Glass
77 mm
14/23 (Centre)