Learn More
Quickfit™ Borosilicate Glass Round Bottom Flask with Septum Side Neck
Designed for semi-micro and micro scale preparations.
Brand: Quickfit FR25/1S/SEP
This item is not returnable.
View return policy
Description
- One vertical centre neck having precision-ground Quickfit™ socket to accept Quickfit™ cone
- One angled side neck designed to take a rubber septum seal to allow small quantities of liquid to be handled easily and safely using a syringe
Specifications
25 mL | |
Designed for semi-micro and micro scale preparations | |
Septum side neck | |
Round |
Borosilicate Glass | |
77 mm | |
14/23 (Centre) |