Learn More
Quickfit™ Borosilicate Glass Round Bottom Flask with Septum Side Neck
Designed for semi-micro and micro scale preparations.
2520.00 SEK - 3905.00 SEK
Specifications
Material Borosilicate Glass
For Use With (Application) Designed for semi-micro and micro scale preparations
Neck Style Septum side neck
Bottom Shape Round
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15577850
View Documents
Quickfit
FR10/1S/SEP
10 mL
3905.00 SEK
Pack of 5
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
15527870
View Documents
Quickfit
FR25/1S/SEP
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • One vertical centrer neck having precision-ground Quickfit™ socket to accept Quickfit™ cone
  • One angled side neck designed to take a rubber septum seal to allow small quantities of liquid to be handled easily and safely using a syringe
Specifications
Specifications
Borosilicate Glass
Designed for semi-micro and micro scale preparations
Septum side neck
Round