missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Round Bottom Flask with Septum Side Neck
Designed for semi-micro and micro scale preparations.
Brand: Quickfit FR10/1S/SEP
3562.50 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- One vertical centre neck having precision-ground Quickfit™ socket to accept Quickfit™ cone
- One angled side neck designed to take a rubber septum seal to allow small quantities of liquid to be handled easily and safely using a syringe
Specifications
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
60 mm | |
14/23 |
10 mL | |
Designed for semi-micro and micro scale preparations | |
Septum side neck | |
Round |