Learn More
(R)-(-)-4-Penten-2-ol, 95%, Thermo Scientific Chemicals
1585.00 SEK - 4915.00 SEK
Chemical Identifiers
CAS 64584-92-5
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD03701536
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYNA-N
Synonym r---4-penten-2-ol, 2r-pent-4-en-2-ol, r-pent-4-en-2-ol, 4-penten-2-ol, 2r, r-4-penten-2-ol, 2r-4-penten-2-ol, 4-penten-2 r-ol, r-2-hydroxy-4-pentene
PubChem CID 6999724
SMILES CC(O)CC=C
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339919
SDS View Documents
Thermo Scientific Alfa Aesar
B25659.03
1 g
1585.00 SEK
1g
11349919
SDS View Documents
Thermo Scientific Alfa Aesar
B25659.06
5 g
4915.00 SEK
5g
Description
Description
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
Solubility
Soluble in water, 4.526 mg/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
(R)-(-)-4-Penten-2-ol, 95%, Thermo Scientific Chemicals