Learn More
Thermo Scientific™ R2 A Agar
Enumerate heterotrophic bacteria from drinking water samples with this ready-to-use medium.
125.00 SEK - 142.00 SEK
Specifications
Description | R2A Agar |
---|---|
Format | 90mm Plate |
Product Type | Agar |
Quantity | 10/Pk. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12932628
|
Thermo Scientific™
PO0659A |
Straw, clear |
125.00 SEK
Pack of 10 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10736793
|
Thermo Scientific™
PO5149A |
Oyster white, transparent |
142.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
Enumerate heterotrophic bacteria from drinking water samples with ready-to-use Thermo Scientific™ Oxoid™ R2A Agar. This nutritionally reduced medium was specially designed to enhance the recovery of stressed and chlorine damaged bacteria from treated water samples.
Use R2A Agar for the microbiological examination of drinking water.
- Aids recovery of stressed and damaged bacteria giving more realistic colony count as compared to traditional media.
- Ready-to-use convenience of prepared media.
Standard methods for the enumeration of heterotrophic bacteria in water have traditionally used nutritionally rich media, such as Plate Count Agar, with incubation at 35°C1. Organisms isolated under these conditions may represent only a small percentage of the bacteria present in the sample2.
R2A Agar developed by Reasoner and Geldreich2 is a nutritionally reduced medium. It was demonstrated that using this medium and incubating for longer at lower temperatures enhanced the recovery of stressed and chlorine damaged bacteria from treated waters resulting in higher, more realistic bacterial counts.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
R2A Agar | |
90mm Plate | |
Agar | |
10/Pk. |
Sécurité et traitement
missing translation for 'shelfLife' : 9 Weeks